Dimethyl 4-Hydroxypyrazole-3,5-dicarboxylate

Supplier: Apollo Scientific

OR471496-1G OR471496-5G
APOSOR471496-1GEA 185 GBP
APOSOR471496-1G APOSOR471496-5G
Dimethyl 4-Hydroxypyrazole-3,5-dicarboxylate
Dimethyl 4-Hydroxypyrazole-3,5-dicarboxylate
Formula: C₇H₈N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD05666011
CAS Number: 23705-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR