N-(2-Hydroxyethyl)-N-methylformamide

Supplier: Apollo Scientific

OR471488-1G OR471488-25G OR471488-5G
APOSOR471488-1GEA 185 GBP
APOSOR471488-1G APOSOR471488-25G APOSOR471488-5G
N-(2-Hydroxyethyl)-N-methylformamide
N-(2-Hydroxyethyl)-N-methylformamide
Formula: C₄H₉NO₂
MW: 103,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16687622
CAS Number: 1590-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR