4-Bromo-2-(1-hydroxyethyl)pyridine
Supplier: Apollo Scientific
OR471467-1G
OR471467-250MG
APOSOR471467-1GEA
726
GBP
APOSOR471467-1G
APOSOR471467-250MG
4-Bromo-2-(1-hydroxyethyl)pyridine
4-Bromo-2-(1-hydroxyethyl)pyridine
Formula:
C₇H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD28131961 CAS Number: 1471260-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...