2-Amino-6-bromo-3,5-dimethylbenzoic acid

Supplier: Apollo Scientific

OR471393-1G OR471393-250MG
APOSOR471393-1GEA 1010 GBP
APOSOR471393-1G APOSOR471393-250MG
2-Amino-6-bromo-3,5-dimethylbenzoic acid
2-Amino-6-bromo-3,5-dimethylbenzoic acid
Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD28977407
CAS Number: 1603580-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR