2-Boc-2,3,3a,4,5,9b-hexahydro-1H-pyrrolo[3,4-c]quinoline

Supplier: Apollo Scientific

OR471310-1G OR471310-250MG
APOSOR471310-1GEA 909 GBP
APOSOR471310-1G APOSOR471310-250MG
2-Boc-2,3,3a,4,5,9b-hexahydro-1H-pyrrolo[3,4-c]quinoline
2-Boc-2,3,3a,4,5,9b-hexahydro-1H-pyrrolo[3,4-c]quinoline
Formula: C₁₆H₂₂N₂O₂
MW: 274,358 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877825
CAS Number: 1194375-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR