4-[(tert-Butylamino)sulphonyl]benzoic acid
Supplier: Apollo Scientific
Synonyms:
4-[(tert-Butylamino)sulphonyl]benzoic acid
OR470988-1G
OR470988-100MG
OR470988-250MG
APOSOR470988-1GEA
899
GBP
APOSOR470988-1G
APOSOR470988-100MG
APOSOR470988-250MG
4-[(tert-Butylamino)sulphonyl]benzoic acid
4-[(tert-Butylamino)sulphonyl]benzoic acid
Formula:
C₁₁H₁₅NO₄S MW: 257,306 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001320 CAS Number: 99987-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...