1-Boc-4-[2-(aminomethyl)phenyl]piperazine
Supplier: Apollo Scientific
OR470438-1G
OR470438-5G
APOSOR470438-1GEA
368
GBP
APOSOR470438-1G
APOSOR470438-5G
1-Boc-4-[2-(aminomethyl)phenyl]piperazine
1-Boc-4-[2-(aminomethyl)phenyl]piperazine
Formula:
C₁₆H₂₅N₃O₂ MW: 291,389 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115066 CAS Number: 174855-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...