4-(Boc-aminoethyloxy)benzonitrile

Supplier: Apollo Scientific

OR470131-5G OR470131-1G
APOSOR470131-5GEA 533 GBP
APOSOR470131-5G APOSOR470131-1G
4-(Boc-aminoethyloxy)benzonitrile
4-(Boc-aminoethyloxy)benzonitrile
Formula: C₁₄H₁₈N₂O₃
MW: 262,304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18070986
CAS Number: 919085-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR