2-(Boc-amino)-3-methylbutylamine

Supplier: Apollo Scientific

OR470055-1G OR470055-5G
APOSOR470055-1GEA 231 GBP
APOSOR470055-1G APOSOR470055-5G
2-(Boc-amino)-3-methylbutylamine
2-(Boc-amino)-3-methylbutylamine
Formula: C₁₀H₂₂N₂O₂
MW: 202,294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828716
CAS Number: 871235-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR