1-tert-Butyl-4-chloro-pyrazolo[3,4-d]pyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR46758-1G
OR46758-250MG
APOSOR46758-1GEA
103
GBP
APOSOR46758-1G
APOSOR46758-250MG
1-tert-Butyl-4-chloro-pyrazolo[3,4-d]pyrimidine
1-tert-Butyl-4-chloro-pyrazolo[3,4-d]pyrimidine
Formula:
C₉H₁₁ClN₄ MW: 210,663 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11104747 CAS Number: 1100365-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...