3-(Aminomethyl)-6-chloropyridazine 95%

Supplier: Apollo Scientific

Synonyms: (6-Chloropyridazin-3-yl)methylamine

OR46079-1G OR46079-500MG
APOSOR46079-1GEA 1240 GBP
APOSOR46079-1G APOSOR46079-500MG
3-(Aminomethyl)-6-chloropyridazine 95%
3-(Aminomethyl)-6-chloropyridazine
Formula: C₅H₆ClN₃
MW: 143,574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698046
CAS Number: 871826-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR