1-(3-Cyclopropylphenyl)sulphonylpiperazine
Supplier: Apollo Scientific
|
Warning
|
OR45227-1G
OR45227-250MG
APOSOR45227-1GEA
629
GBP
APOSOR45227-1G
APOSOR45227-250MG
1-(3-Cyclopropylphenyl)sulphonylpiperazine
1-(3-Cyclopropylphenyl)sulphonylpiperazine
Formula:
C₁₃H₁₈N₂O₂S MW: 266,359 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32219474 CAS Number: 2416262-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...