N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride 97%

Supplier: Apollo Scientific

OR452106-1G OR452106-250MG OR452106-500MG
APOSOR452106-1GEA 787 GBP
APOSOR452106-1G APOSOR452106-250MG APOSOR452106-500MG
N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride 97%
N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride
Formula: C₇H₁₄N₂O·ClH
MW: 178,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17214731
CAS Number: 1211573-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR