1-Methyl-3-pyrrolidineacetic acid hydrochloride 97%
Supplier: Apollo Scientific
Synonyms:
2-(1-Methylpyrrolidin-3-yl)acetic acid hydrochloride
OR452105-1G
OR452105-250MG
OR452105-500MG
APOSOR452105-1GEA
787
GBP
APOSOR452105-1G
APOSOR452105-250MG
APOSOR452105-500MG
1-Methyl-3-pyrrolidineacetic acid hydrochloride 97%
1-Methyl-3-pyrrolidineacetic acid hydrochloride
Formula:
C₇H₁₃NO₂·ClH MW: 179,645 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23135758 CAS Number: 1071634-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...