(R)-2-(3-Chlorophenyl)glycine 97%

Supplier: Apollo Scientific

Synonyms: (R)-2-Amino-2-(3-chlorophenyl)acetic acid, H-D-Phg(3-Cl)-OH

OR452013-1G OR452013-500MG OR452013-5G
APOSOR452013-1GEA 237 GBP
APOSOR452013-1G APOSOR452013-500MG APOSOR452013-5G
(R)-2-(3-Chlorophenyl)glycine 97%
(R)-2-(3-Chlorophenyl)glycine
Formula: C₈H₈ClNO₂
MW: 185,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371715
CAS Number: 25698-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR