4-(2-Bromoethyl)benzonitrile

Supplier: Apollo Scientific
Warning

OR45144-1G OR45144-250MG OR45144-5G
APOSOR45144-1GEA 24.6 GBP
APOSOR45144-1G APOSOR45144-250MG APOSOR45144-5G
4-(2-Bromoethyl)benzonitrile
4-(2-Bromoethyl)benzonitrile
Formula: C₉H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD12025077
CAS Number: 72054-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR