8-Chloro-3,4-dihydro-1H-quinolin-2-one

Supplier: Apollo Scientific

OR450049-100MG OR450049-1G OR450049-250MG
APOSOR450049-100MGEA 109 GBP
APOSOR450049-100MG APOSOR450049-1G APOSOR450049-250MG
8-Chloro-3,4-dihydro-1H-quinolin-2-one
8-Chloro-3,4-dihydro-1H-quinolin-2-one
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD19276419
CAS Number: 83229-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR