Ethyl 2,4-dihydroxy-6-pentylbenzoate

Supplier: Apollo Scientific

OR450019-1G OR450019-5G
APOSOR450019-1GEA 74.8 GBP
APOSOR450019-1G APOSOR450019-5G
Ethyl 2,4-dihydroxy-6-pentylbenzoate
Ethyl 2,4-dihydroxy-6-pentylbenzoate
Formula: C₁₄H₂₀O₄
MW: 252,306 g/mol
Melting Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD11618127
CAS Number: 38862-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR