2-(4-Methoxybenzyloxy)-5-chlorophenylacetic acid 95%

Supplier: Apollo Scientific
Warning

OR42280-1G OR42280-250MG
APOSOR42280-1GEA 14.5 GBP
APOSOR42280-1G APOSOR42280-250MG
2-(4-Methoxybenzyloxy)-5-chlorophenylacetic acid 95%
2-(4-Methoxybenzyloxy)-5-chlorophenylacetic acid
Formula: C₁₆H₁₅ClO₄
MW: 306,741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31726534
CAS Number: 2367002-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR