2-Bromo-4-cyclopropylaniline 96%

Supplier: Apollo Scientific
Warning

OR40777-1G OR40777-250MG OR40777-5G
APOSOR40777-1GEA 446 GBP
APOSOR40777-1G APOSOR40777-250MG APOSOR40777-5G
2-Bromo-4-cyclopropylaniline 96%
2-Bromo-4-cyclopropylaniline
Formula: C₉H₁₀BrN
MW: 212,086 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21099997
CAS Number: 1353857-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR