2-Chloro-6,7-dihydro-4-methyl-5H-cyclopenta[d]pyrimidine 95%

Supplier: Apollo Scientific
Warning

OR40569-1G OR40569-500MG
APOSOR40569-1GEA 446 GBP
APOSOR40569-1G APOSOR40569-500MG
2-Chloro-6,7-dihydro-4-methyl-5H-cyclopenta[d]pyrimidine 95%
2-Chloro-6,7-dihydro-4-methyl-5H-cyclopenta[d]pyrimidine
Formula: C₈H₉ClN₂
MW: 168,623 g/mol
Melting Pt: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD14584769
CAS Number: 83939-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR