Ethyl 3,6-dichloropyridine-2-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3,6-dichloropicolinate, 3,6-Dichloro-2-(ethoxycarbonyl)pyridine

OR40369-1G OR40369-5G
APOSOR40369-1GEA 91.8 GBP
APOSOR40369-1G APOSOR40369-5G
Ethyl 3,6-dichloropyridine-2-carboxylate
Ethyl 3,6-dichloropyridine-2-carboxylate
Formula: C₈H₇Cl₂NO₂
MW: 220,053 g/mol
Boiling Pt: 94 °C (0,5 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD14698221
CAS Number: 253440-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR