2-Bromo-6-chlorostyrene 95%

Supplier: Apollo Scientific
Warning

OR400920-1G OR400920-250MG
APOSOR400920-1GEA 511 GBP
APOSOR400920-1G APOSOR400920-250MG
2-Bromo-6-chlorostyrene 95%
2-Bromo-6-chlorostyrene
Formula: C₈H₆BrCl
MW: 217.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31561244
CAS Number: 2228365-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR