5-Bromo-2-nitrobenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Bromo-2-(bromomethyl)-1-nitrobenzene
OR400606-1G
OR400606-250MG
OR400606-5G
APOSOR400606-1GEA
73.3
GBP
APOSOR400606-1G
APOSOR400606-250MG
APOSOR400606-5G
5-Bromo-2-nitrobenzyl bromide
5-Bromo-2-nitrobenzyl bromide
Formula:
C₇H₅Br₂NO₂ MW: 294.928 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848689 CAS Number: 35287-42-4 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...