2-Bromo-3-iodobenzyl alcohol

Supplier: Apollo Scientific
Warning

Synonyms: (2-Bromo-3-iodophenyl)methanol

OR400516-1G OR400516-5G
APOSOR400516-1GEA 218 GBP
APOSOR400516-1G APOSOR400516-5G
2-Bromo-3-iodobenzyl alcohol
2-Bromo-3-iodobenzyl alcohol
Formula: C₇H₆BrIO
MW: 312,93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520410
CAS Number: 1261644-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR