3-Bromo-5-methoxybenzene-1,2-diamine 95%

Supplier: Apollo Scientific
Warning

OR400424-1G OR400424-250MG OR400424-5G
APOSOR400424-1GEA 123 GBP
APOSOR400424-1G APOSOR400424-250MG APOSOR400424-5G
3-Bromo-5-methoxybenzene-1,2-diamine 95%
3-Bromo-5-methoxybenzene-1,2-diamine
Formula: C₇H₉BrN₂O
MW: 217,063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144648
CAS Number: 98546-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR