4-Bromo-3-methoxybenzaldehyde 98%

Supplier: Apollo Scientific
Warning

OR400178-1G OR400178-5G OR400178-10G
APOSOR400178-1GEA 48.6 GBP
APOSOR400178-1G APOSOR400178-5G APOSOR400178-10G
4-Bromo-3-methoxybenzaldehyde 98%
4-Bromo-3-methoxybenzaldehyde
Formula: C₈H₇BrO₂
MW: 215,044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846303
CAS Number: 43192-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR