4-Piperazinylphenylboronic acid, pinacol ester 95%

Supplier: Apollo Scientific
Danger

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine

OR361543-1G OR361543-250MG
APOSOR361543-1GEA 194 GBP
APOSOR361543-1G APOSOR361543-250MG
4-Piperazinylphenylboronic acid, pinacol ester 95%
4-Piperazinylphenylboronic acid, pinacol ester
Formula: C₁₆H₂₅BN₂O₂
MW: 288,193 g/mol
Melting Pt: 135…143 °C
Storage Temperature: Ambient
MDL Number: MFCD06795649
CAS Number: 912369-50-7
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR