2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 95%

Supplier: Apollo Scientific
Danger

Synonyms: N-Methyl-1-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine

OR361411-1G OR361411-250MG
APOSOR361411-1GEA 449 GBP
APOSOR361411-1G APOSOR361411-250MG
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 95%
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester
Formula: C₁₄H₂₂BNO₂
MW: 247,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855967
CAS Number: 1150271-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR