4-Bromo-2-methylphenylboronic acid 98%

Supplier: Apollo Scientific

OR360799-1G OR360799-250MG
APOSOR360799-1GEA 0 GBP
APOSOR360799-1G APOSOR360799-250MG
4-Bromo-2-methylphenylboronic acid 98%
4-Bromo-2-methylphenylboronic acid
Formula: C₇H₈BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD16660270
CAS Number: 221006-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR