1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%

Supplier: Apollo Scientific

Synonyms: 1-(Cyclopropylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR360142-1G OR360142-250MG
APOSOR360142-1GEA 149 GBP
APOSOR360142-1G APOSOR360142-250MG
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester
Formula: C₁₃H₂₁BN₂O₂
MW: 248,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15474887
CAS Number: 1000801-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR