6-Methylpyridine-3-sulphonyl chloride 95+%

Supplier: Apollo Scientific

OR346514-1G OR346514-5G
APOSOR346514-1GEA 239 GBP
APOSOR346514-1G APOSOR346514-5G
6-Methylpyridine-3-sulphonyl chloride 95+%
6-Methylpyridine-3-sulphonyl chloride
Formula: C₆H₆ClNO₂S
MW: 191,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11868981
CAS Number: 478264-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR