3-Bromo-2-methylbenzenesulphonyl chloride 95+%
Supplier: Apollo Scientific
Synonyms:
3-Bromo-2-methylbenzene-1-sulphonyl chloride
OR346500-1G
OR346500-5G
APOSOR346500-1GEA
257
GBP
APOSOR346500-1G
APOSOR346500-5G
3-Bromo-2-methylbenzenesulphonyl chloride 95+%
3-Bromo-2-methylbenzenesulphonyl chloride
Formula:
C₇H₆BrClO₂S MW: 269,543 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782194 CAS Number: 886501-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...