4-(3-Methylisothiazol-5-yl)benzaldehyde 95+%

Supplier: Apollo Scientific

OR346461-1G OR346461-500MG
APOSOR346461-1GEA 334 GBP
APOSOR346461-1G APOSOR346461-500MG
4-(3-Methylisothiazol-5-yl)benzaldehyde 95+%
4-(3-Methylisothiazol-5-yl)benzaldehyde
Formula: C₁₁H₉NOS
MW: 203,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573494
CAS Number: 1401521-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR