4-Methylbenzimidic acid methyl ester hydrochloride 95+%

Supplier: Apollo Scientific

OR346452-1G OR346452-500MG OR346452-5G
APOSOR346452-1GEA 239 GBP
APOSOR346452-1G APOSOR346452-500MG APOSOR346452-5G
4-Methylbenzimidic acid methyl ester hydrochloride 95+%
4-Methylbenzimidic acid methyl ester hydrochloride
Formula: C₉H₁₁NO·ClH
MW: 185,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981473
CAS Number: 39739-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR