2-(4,4-Dimethyl-4,5-dihydro-oxazol-2-yl)-3-methylpyridine 95+%
Supplier: Apollo Scientific
OR346427-1G
OR346427-500MG
OR346427-5G
APOSOR346427-1GEA
239
GBP
APOSOR346427-1G
APOSOR346427-500MG
APOSOR346427-5G
2-(4,4-Dimethyl-4,5-dihydro-oxazol-2-yl)-3-methylpyridine 95+%
2-(4,4-Dimethyl-4,5-dihydro-oxazol-2-yl)-3-methylpyridine
Formula:
C₁₁H₁₄N₂O MW: 190,242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18259374 CAS Number: 525602-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...