3-(Chloromethyl)-5-(2-chlorophenyl)-[1,2,4]oxadiazole 95+%
Supplier: Apollo Scientific
OR346406-1G
OR346406-500MG
OR346406-5G
APOSOR346406-1GEA
239
GBP
APOSOR346406-1G
APOSOR346406-500MG
APOSOR346406-5G
3-(Chloromethyl)-5-(2-chlorophenyl)-[1,2,4]oxadiazole 95+%
3-(Chloromethyl)-5-(2-chlorophenyl)-[1,2,4]oxadiazole
Formula:
C₉H₆Cl₂N₂O MW: 229,063 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08444943 CAS Number: 110704-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...