1-m-Tolyl-1H-pyrazole-4-carboxaldehyde 95+%

Supplier: Apollo Scientific

OR346396-1G OR346396-5G
APOSOR346396-1GEA 182 GBP
APOSOR346396-1G APOSOR346396-5G
1-m-Tolyl-1H-pyrazole-4-carboxaldehyde 95+%
1-m-Tolyl-1H-pyrazole-4-carboxaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03780742
CAS Number: 400876-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR