5-(Chloromethyl)-4-methyl-[1,2,3]thiadiazole 95+%

Supplier: Apollo Scientific

OR346389-1G OR346389-5G
APOSOR346389-1GEA 125 GBP
APOSOR346389-1G APOSOR346389-5G
5-(Chloromethyl)-4-methyl-[1,2,3]thiadiazole 95+%
5-(Chloromethyl)-4-methyl-[1,2,3]thiadiazole
Formula: C₄H₅ClN₂S
MW: 148,614 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10686610
CAS Number: 82172-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR