(5-Methyl-3-phenylpyrazol-1-yl)methanol 95+%

Supplier: Apollo Scientific

OR346385-1G OR346385-5G
APOSOR346385-1GEA 182 GBP
APOSOR346385-1G APOSOR346385-5G
(5-Methyl-3-phenylpyrazol-1-yl)methanol 95+%
(5-Methyl-3-phenylpyrazol-1-yl)methanol
Formula: C₁₁H₁₂N₂O
MW: 188,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227224
CAS Number: 369401-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR