2-(2-Chloroethylamino)-4,6-dimethylnicotinonitrile 95+%
Supplier: Apollo Scientific
OR346378-1G
OR346378-5G
APOSOR346378-1GEA
151
GBP
APOSOR346378-1G
APOSOR346378-5G
2-(2-Chloroethylamino)-4,6-dimethylnicotinonitrile 95+%
2-(2-Chloroethylamino)-4,6-dimethylnicotinonitrile
Formula:
C₁₀H₁₂ClN₃ MW: 209,675 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568281 CAS Number: 1053659-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...