2-(2-Chloroethoxy)-4,6-dimethylnicotinonitrile 95+%

Supplier: Apollo Scientific

OR346377-1G OR346377-5G
APOSOR346377-1GEA 151 GBP
APOSOR346377-1G APOSOR346377-5G
2-(2-Chloroethoxy)-4,6-dimethylnicotinonitrile 95+%
2-(2-Chloroethoxy)-4,6-dimethylnicotinonitrile
Formula: C₁₀H₁₁ClN₂O
MW: 210,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568279
CAS Number: 1053658-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR