1-Benzyl-1H-pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
OR346360-1G
OR346360-5G
APOSOR346360-1GEA
108
GBP
APOSOR346360-1G
APOSOR346360-5G
1-Benzyl-1H-pyrazole-4-carboxylic acid
1-Benzyl-1H-pyrazole-4-carboxylic acid
Formula:
C₁₁H₁₀N₂O₂ MW: 202,209 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864591 CAS Number: 401647-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...