2-Amino-6-bromo-3-methoxypyridine 97%

Supplier: Apollo Scientific

Synonyms: 6-Bromo-3-methoxypyridin-2-amine

OR345483-1G OR345483-250MG OR345483-5G
APOSOR345483-1GEA 144 GBP
APOSOR345483-1G APOSOR345483-250MG APOSOR345483-5G
2-Amino-6-bromo-3-methoxypyridine 97%
2-Amino-6-bromo-3-methoxypyridine
Formula: C₆H₇BrN₂O
MW: 203.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15142726
CAS Number: 916737-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR