Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate 97%

Supplier: Apollo Scientific

OR33611-1G OR33611-5G
APOSOR33611-1GEA 269 GBP
APOSOR33611-1G APOSOR33611-5G
Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate 97%
Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate
Formula: C₁₂H₁₁Cl₂NO₃
MW: 288,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28016143
CAS Number: 1598370-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR