4-Acetoxy-2-bromo-5-methoxybenzaldehyde 98%

Supplier: Apollo Scientific
Warning

OR3346-1G OR3346-5G OR3346-25G
APOSOR3346-1GEA 38.1 GBP
APOSOR3346-1G APOSOR3346-5G APOSOR3346-25G
4-Acetoxy-2-bromo-5-methoxybenzaldehyde 98%
4-Acetoxy-2-bromo-5-methoxybenzaldehyde
Formula: C₁₀H₉BrO₄
MW: 273,08 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00452230
CAS Number: 52783-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR