1-(Dimethylamino)-4-phenoxypent-1-en-3-one 95%

Supplier: Apollo Scientific

OR33327-1G OR33327-5G
APOSOR33327-1GEA 211 GBP
APOSOR33327-1G APOSOR33327-5G
1-(Dimethylamino)-4-phenoxypent-1-en-3-one 95%
1-(Dimethylamino)-4-phenoxypent-1-en-3-one
Formula: C₁₃H₁₇NO₂
MW: 219,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00139117
CAS Number: 338976-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR