N'-(6-Chloropyridin-2-yl)acetohydrazide 95%

Supplier: Apollo Scientific

OR33021-1G OR33021-5G
APOSOR33021-1GEA 139 GBP
APOSOR33021-1G APOSOR33021-5G
N'-(6-Chloropyridin-2-yl)acetohydrazide 95%
N'-(6-Chloropyridin-2-yl)acetohydrazide
Formula: C₇H₈ClN₃O
MW: 185,611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02187335
CAS Number: 66999-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR