1,3-Bis(morpholin-4-yl)propane-1,3-dione 95%
Supplier: Apollo Scientific
OR32967-1G
OR32967-5G
APOSOR32967-1GEA
112
GBP
APOSOR32967-1G
APOSOR32967-5G
1,3-Bis(morpholin-4-yl)propane-1,3-dione 95%
1,3-Bis(morpholin-4-yl)propane-1,3-dione
Formula:
C₁₁H₁₈N₂O₄ MW: 242,272 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023355 CAS Number: 10256-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...