4-Oxo-1-(propan-2-yl)-1,4-dihydroquinoline-3-carboxylic acid 97%
Supplier: Apollo Scientific
OR32639-1G
OR32639-500MG
APOSOR32639-1GEA
576
GBP
APOSOR32639-1G
APOSOR32639-500MG
4-Oxo-1-(propan-2-yl)-1,4-dihydroquinoline-3-carboxylic acid 97%
4-Oxo-1-(propan-2-yl)-1,4-dihydroquinoline-3-carboxylic acid
Formula:
C₁₃H₁₃NO₃ MW: 231,247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01315709 CAS Number: 53977-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...